missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SEPW1 Protein
A cDNA sequence encoding the SEPW1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | SEPW1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953609
|
enQuireBio™
QP13452-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943609
|
enQuireBio™
QP13452-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933609
|
enQuireBio™
QP13452-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SEPW1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SEPW1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALAVRV VYCGACGYKS KYLQLKKKLE DEFPGRLDIC GEGTPQATGF FEVMVAGKLI HSKKKGDGYV DTESKFLKLV AAIKAALAQG | |
Greater than 95.0% as determined by SDS-PAGE. |