missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Septin-7 Protein
A DNA sequence encoding the Homo sapiens (Human) Septin-7, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001779.3 |
---|---|
Gene ID (Entrez) | 989 |
Name | Septin-7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049510
|
enQuireBio™
QP7576-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089510
|
enQuireBio™
QP7576-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059510
|
enQuireBio™
QP7576-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079510
|
enQuireBio™
QP7576-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099510
|
enQuireBio™
QP7576-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069510
|
enQuireBio™
QP7576-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001779.3 | |
Septin-7 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
989 | |
Research Use Only | |
42985 | |
Recombinant Protein | |
Yeast | |
SVSARSAAAE ERSVNSSTMV AQQKNLEGYV GFANLPNQVY RKSVKRGFEF TLMVVGESGL GKSTLINSLF LTDLYSPEYP GPSHRIKKTV QVEQSKVLIK EGGVQLLLTI VDTPGFGDAV DNSNCWQPVI DYIDSKFEDY LNAESRVNRR QMPDNRVQCC LYFIAPSGHG LKPLDIEFMK RLHEKVNIIP LIAKADTLTP EECQQFKKQI MKEIQEHKIK IYEFPETDDE EENKLVKKIK DRLPLAVVGS NTIIEVNGKR VRGRQYPWGV AEVENGEHCD FTILRNMLIR THMQDLKDVT NNVHYENYRS RKLAAVTYNG VDNNKNKGQL TKSPLAQMEE ERREHVAKMK KMEMEMEQVF EMKVKEKVQK LKDSEAELQR RHEQMKKNLE AQHKELEEKR RQFEDEKANW EAQQRILEQQ NSSRTLEKNK KKGKIF | |
Greater than 90% as determined by SDS-PAGE. |