missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SEC22B Protein
A cDNA sequence encoding the SEC22B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13446-5ug
Additional Details : Weight : 0.01000kg
Specifications
SEC22B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSM LPLAAS MQEDEQSGRD LQQYQSQAKQ LFRKLNEQSP TRCTLEAGAM TFHYIIEQGV CYLVLCEAAF PKKLAFAYLE DLHSEFDEQH GKKVPTVSRP YSFIEFDTFI QKTKKLYIDS RARRNLGSIN TELQDVQRIM VANIEEVLQR GEALSALDSK ANNLSSLSKK YRQDAKYLNM RSTYA | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SEC22B solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 0.15M NaCl and 10% glycerol. |