missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SDF2 Protein

A DNA sequence encoding the Homo sapiens (Human) SDF2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6657-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084860

  • £384.00 / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_008854.2
SDF2 Protein
Research Use Only
SDF2
Recombinant Protein
E. coli
SSLGVVTCGS VVKLLNTRHN VRLHSHDVRY GSGSGQQSVT GVTSVDDSNS YWRIRGKSAT VCERGTPIKC GQPIRLTHVN TGRNLHSHHF TSPLSGNQEV SAFGEEGEGD YLDDWTVLCN GPYWVRDGEV RFKHSSTEVL LSVTGEQYGR PISGQKEVHG MAQPSQNNYW KAMEGIFMKP SELLKAEAHH AEL
Greater than 90% as determined by SDS-PAGE.
6388
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers