missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SDCBP Protein

A cDNA sequence encoding the SDCBP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13439-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943569

  • £2622.00 / 1mg

Estimated Shipment
30-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SDCBP Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SDCBP protein solution (0.5 mg/1ml) is formulated in 20 mM Tris-HCl Buffer (pH 8.0), 100 mM NaCl, and 40% Glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSLYPSLEDL KVDKVIQAQT AFSANPANPA ILSEASAPIP HDGNLYPRLY PELSQYMGLS LNEEEIRANV AVVSGAPLQG QLVARPSSIN YMVAPVTGND VGIRRAEIKQ GIREVILCKD QDGKIGLRLK SIDNGIFVQL VQANSPASLV GLRFGDQVLQ INGENCAGWS SDKAHKVLKQ AFGEKITMTI RDRPFERTIT MHKDSTGHVG FIFKNGKITS IVKDSSAARN GLLTEHNICE INGQNVIGLK
Videos
SDS
Documents