missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCAN domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) SCAN domain-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6651-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057642.1 | |
SCAN domain-containing protein 1 | |
Research Use Only | |
SCAND1 | |
Recombinant Protein | |
E. coli | |
MAATEPILAA TGSPAAVPPE KLEGAGSSSA PERNCVGSSL PEASPPAPEP SSPNAAVPEA IPTPRAAASA ALELPLGPAP VSVAPQAEAE ARSTPGPAGS RLGPETFRQR FRQFRYQDAA GPREAFRQLR ELSRQWLRPD IRTKEQIVEM LVQEQLLAIL PEAARARRIR RRTDVRITG | |
Greater than 80% as determined by SDS-PAGE. |
51282 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |