missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAMSN1 Protein
A cDNA sequence encoding the SAMSN1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | SAMSN1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953489
|
enQuireBio™
QP13412-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933489
|
enQuireBio™
QP13412-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943489
|
enQuireBio™
QP13412-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SAMSN1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SAMSN1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol 0.1M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLKRKPS NVSEKEKHQK PKRSSSFGNF DRFRNNSLSK PDDSTEAHEG DPTNGSGEQS KTSNNGGGLG KKMRAISWTM KKKVGKKYIK ALSEEKDEED GENAHPYRNS DPVIGTHTEK VSLKASDSMD SLYSGQSSSS GITSCSDGTS NRDSFRLDDD GPYSGPFCGR ARVHTDFTPS PYDTDSLKIK KGDIIDIICK TPMGMWTGML NNKVGNFKFI YVDVISEEEA APKKIKANRR SNSKKSKTLQ EFLERIHLQE YTSTLLLNGY ETLEDLKDIK ESHLIELNIE NPDDRRRLLS AAENFLEEEI IQEQENEPEP LSLSSDISLN KSQLDDCPRD SGCYISSGNS DNGKEDLESE NLSDMVHKII ITEPSD | |
Greater than 80.0% as determined by SDS-PAGE. |