missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAA Protein
A cDNA sequence encoding the SAA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10860-50ug
Additional Details : Weight : 0.01000kg
Specifications
SAA Protein | |
Research Use Only | |
The biological activity determined by its ability to down-regulate lipid biosynthesis in aortic smooth muscle cells. The effective concentration was found to be 4microm. | |
Human | |
His | |
Human SAA was lyophilized from a concentrated (1 mg/ml) solution in 20mM Tris-HCl, pH 9.0 and 150mM NaCl. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
RSFFSFLGEA FDGARDMWRA YSDMREANYI GSDKYFHARG NYDAAKRGPG GVWAAEAISN ARENIQRFFG RGAEDSLADQ AANEWGRSGK DPNHFRPAGL PEKY | |
Greater than 98.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |