missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100Z Protein
A cDNA sequence encoding the S100Z was constructed and used to recombinantly synthesize the protein.
£99.15 - £2490.00
Specifications
Name | S100Z Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963479
|
enQuireBio™
QP13409-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953479
|
enQuireBio™
QP13409-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943479
|
enQuireBio™
QP13409-1MG |
1 mg |
£2490.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
S100Z Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
S100Z Human solution (1 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT, 1mM EDTA, 50mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPTQLEMAMD TMIRIFHRYS GKERKRFKLS KGELKLLLQR ELTEFLSCQK ETQLVDKIVQ DLDANKDNEV DFNEFVVMVA ALTVACNDYF VEQLKKKGK | |
Greater than 95.0% as determined by SDS-PAGE. |