missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RYBP Protein

A cDNA sequence encoding the RYBP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13379-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963379

  • £1324.47 / 100µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

23429
Greater than 85% as determined by SDS-PAGE.
Research Use Only
RYBP
Human
His
The RYBP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol.
RYBP Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMTMGDK KSPTRPKRQA KPAADEGFWD CSVCTFRNSA EAFKCSICDV RKGTSTRKPR INSQLVAQQV AQQYATPPPP KKEKKEKVEK QDKEKPEKDK EISPSVTKKN TNKKTKPKSD ILKDPPSEAN SIQSANATTK TSETNHTSRP RLKNVDRSTA QQLAVTVGNV TVIITDFKEK TRSSSTSSST VTSSAGSEQQ NQSSSGSEST DKGSSRSSTP KGDMSAVNDE SF
Videos
SDS
Documents
Certificates
Special Offers

Special Offers