missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RuvB-like 2 Protein
A DNA sequence encoding the Homo sapiens (Human) RuvB-like 2, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001308120.1 |
---|---|
Gene ID (Entrez) | 10856 |
Name | RuvB-like 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053841
|
enQuireBio™
QP8295-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073841
|
enQuireBio™
QP8295-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063841
|
enQuireBio™
QP8295-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083841
|
enQuireBio™
QP8295-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093841
|
enQuireBio™
QP8295-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003851
|
enQuireBio™
QP8295-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001308120.1 | |
RuvB-like 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10856 | |
Research Use Only | |
RUVBL2 | |
Recombinant Protein | |
E. coli | |
ATVTATTKVP EIRDVTRIER IGAHSHIRGL GLDDALEPRQ ASQGMVGQLA ARRAAGVVLE MIREGKIAGR AVLIAGQPGT GKTAIAMGMA QALGPDTPFT AIAGSEIFSL EMSKTEALTQ AFRRSIGVRI KEETEIIEGE VVEIQIDRPA TGTGSKVGKL TLKTTEMETI YDLGTKMIES LTKDKVQAGD VITIDKATGK ISKLGRSFTR ARDYDAMGSQ TKFVQCPDGE LQKRKEVVHT VSLHEIDVIN SRTQGFLALF SGDTGEIKSE VREQINAKVA EWREEGKAEI IPGVLFIDEV HMLDIESFSF LNRALESDMA PVLIMATNRG ITRIRGTSYQ SPHGIPIDLL DRLLIVSTTP YSEKDTKQIL RIRCEEEDVE MSEDAYTVLT RIGLETSLRY AIQLITAASL VCRKRKGTEV QVDDIKRVYS LFLDESRSTQ YMKEYQDAFL FNELKGETMD TS | |
Greater than 90% as determined by SDS-PAGE. |