missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RQCD1 Protein
A cDNA sequence encoding the RQCD1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | RQCD1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943329
|
enQuireBio™
QP13361-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933329
|
enQuireBio™
QP13361-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923329
|
enQuireBio™
QP13361-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RQCD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RQCD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MHSLATAAPV PTTLAQVDRE KIYQWINELS SPETRENALL ELSKKRESVP DLAPMLWHSF GTIAALLQEI VNIYPSINPP TLTAHQSNRV CNALALLQCV ASHPETRSAF LAAHIPLFLY PFLHTVSKTR PFEYLRLTSL GVIGALVKTD EQEVINFLLT TEIIPLCLRI MESGSELSKT VATFILQKIL LDDTGLAYIC QTYERFSHVA MILGKMVLQL SKEPSARLLK HVVRCYLRLS DNPRAREALR QCLPDQLKDT TFAQVLKDDT TTKRWLAQLV KNLQEGQVTD PRGIPLPPQ | |
Greater than 95.0% as determined by SDS-PAGE. |