missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribose-phosphate pyrophosphokinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribose-phosphate pyrophosphokinase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6535-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001191331.1 | |
Ribose-phosphate pyrophosphokinase 1 Protein | |
Research Use Only | |
PRPS1 | |
Recombinant Protein | |
E. coli | |
PNIKIFSGSS HQDLSQKIAD RLGLELGKVV TKKFSNQETC VEIGESVRGE DVYIVQSGCG EINDNLMELL IMINACKIAS ASRVTAVIPC FPYARQDKKD KSRAPISAKL VANMLSVAGA DHIITMDLHA SQIQGFFDIP VDNLYAEPAV LKWIRENISE WRNCTIVSPD AGGAKRVTSI ADRLNVDFAL IHKERKKANE VDRMVLVGDV KDRVAILVDD MADTCGTICH AADKLLSAGA TRVYAILTHG IFSGPAISRI NNACFEAVVV TNTIPQEDKM KHCSKIQVID ISMILAEAIR RTHNGESVSY LFSHVPL | |
Greater than 90% as determined by SDS-PAGE. |
5631 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |