missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RHOQ Protein
A cDNA sequence encoding the RHOQ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13319-2ug
Additional Details : Weight : 0.01000kg
Specifications
RHOQ Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAHGPGA LMLKCVVVGD GAVGKTCLLM SYANDAFPEE YVPTVFDHYA VSVTVGGKQY LLGLYDTAGQ EDYDRLRPLS YPMTDVFLIC FSVVNPASFQ NVKEEWVPEL KEYAPNVPFL LIGTQIDLRD DPKTLARLND MKEKPICVEQ GQKLAKEIGA CCYVECSALT QKGLKTVFDE AIIAILTPKK HTVKKRIGSR CINCC | |
Greater than 85% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RHOQ protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |