missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RGS4 Protein

A cDNA sequence encoding the RGS4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13312-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953169

  • £99.15 / 2µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RGS4 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MCKGLAGLPA SCLRSAKDMK HRLGFLLQKS DSCEHNSSHN KKDKVVICQR VSQEEVKKWA ESLENLISHE CGLAAFKAFL KSEYSEENID FWISCEEYKK IKSPSKLSPK AKKIYNEFIS VQATKEVNLD SCTREETSRN MLEPTITCFD EAQKKIFNLM EKDSYRRFLK SRFYLDLVNP SSCGAEKQKG AKSSADCASL VPQCA
Greater than 90% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RGS4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 10% glycerol and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers