missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RGS1 Protein

A cDNA sequence encoding the RGS1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13306-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963149

  • £151.37 / 20µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RGS1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RGS1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMRAAAI STPKLDKMPG MFFSANPKEL KGTTHSLLDD KMQKRRPKTF GMDMKAYLRS MIPHLESGMK SSKSKDVLSA AEVMQWSQSL EKLLANQTGQ NVFGSFLKSE FSEENIEFWL ACEDYKKTES DLLPCKAEEI YKAFVHSDAA KQINIDFRTR ESTAKKIKAP TPTCFDEAQK VIYTLMEKDS YPRFLKSDIY LNLLNDLQAN SLK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers