missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Renin receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Renin receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5698-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957099

  • £246.00 / 50µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005756.2
Renin receptor Protein
Research Use Only
ATP6AP2
Recombinant Protein
E. coli
NEFSILKSPG SVVFRNGNWP IPGERIPDVA ALSMGFSVKE DLSWPGLAVG NLFHRPRATV MVMVKGVNKL ALPPGSVISY PLENAVPFSL DSVANSIHSL FSEETPVVLQ LAPSEERVYM VGKANSVFED LSVTLRQLRN RLFQENSVLS SLPLNSLSRN NEVDLLFLSE LQVLHDISSL LSRHKHLAKD HSPDLYSLEL AGLDEIGKRY GEDSEQFRDA SKILVDALQK FADDMYSLYG GNAVVELVTV KSFDTSLIRK TRTILEAKQA KNPASPYNLA YKYNFEYSVV FNMVLWIMIA LALAVIITSY NIWNMDPGYD SIIYRMTNQK IRMD
Greater than 90% as determined by SDS-PAGE.
10159
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers