missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Renin receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Renin receptor, was expressed in the hosts and tags indicated.
£99.15 - £1780.00
Specifications
Accession Number | NP_005756.2 |
---|---|
Gene ID (Entrez) | 10159 |
Name | Renin receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15917099
|
enQuireBio™
QP5698-EC-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957099
|
enQuireBio™
QP5698-EC-50UG |
50 μg |
£246.00
50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907099
|
enQuireBio™
QP5698-EC-100UG |
100 μg |
£393.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937099
|
enQuireBio™
QP5698-EC-200UG |
200 μg |
£635.00
200µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947099
|
enQuireBio™
QP5698-EC-500UG |
500 μg |
£1095.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927099
|
enQuireBio™
QP5698-EC-1MG |
1 mg |
£1780.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_005756.2 | |
Renin receptor Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10159 | |
Research Use Only | |
ATP6AP2 | |
Recombinant Protein | |
E. coli | |
NEFSILKSPG SVVFRNGNWP IPGERIPDVA ALSMGFSVKE DLSWPGLAVG NLFHRPRATV MVMVKGVNKL ALPPGSVISY PLENAVPFSL DSVANSIHSL FSEETPVVLQ LAPSEERVYM VGKANSVFED LSVTLRQLRN RLFQENSVLS SLPLNSLSRN NEVDLLFLSE LQVLHDISSL LSRHKHLAKD HSPDLYSLEL AGLDEIGKRY GEDSEQFRDA SKILVDALQK FADDMYSLYG GNAVVELVTV KSFDTSLIRK TRTILEAKQA KNPASPYNLA YKYNFEYSVV FNMVLWIMIA LALAVIITSY NIWNMDPGYD SIIYRMTNQK IRMD | |
Greater than 90% as determined by SDS-PAGE. |