missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human REN Protein
A cDNA sequence encoding the REN was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | REN Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943119
|
enQuireBio™
QP13296-HIS-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953119
|
enQuireBio™
QP13296-HIS-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923119
|
enQuireBio™
QP13296-HIS-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933119
|
enQuireBio™
QP13296-HIS-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963119
|
enQuireBio™
QP13296-HIS-EC-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
REN Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The REN solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLTLGN TTSSVILTNY MDTQYYGEIG IGTPPQTFKV VFDTGSSNVW VPSSKCSRLY TACVYHKLFD ASDSSSYKHN GTELTLRYST GTVSGFLSQD IITVGGITVT QMFGEVTEMP ALPFMLAEFD GVVGMGFIEQ AIGRVTPIFD NIISQGVLKE DVFSFYYNRD SENSQSLGGQ IVLGGSDPQH YEGNFHYINL IKTGVWQIQM KGVSVGSSTL LCEDGCLALV DTGASYISGS TSSIEKLMEA LGAKKRLFDY VVKCNEGPTL PDISFHLGGK EYTLTSADYV FQESYSSKKL CTLAIHAMDI PPPTGPTWAL GATFIRKFYT EFDRRNNRIG FALAR | |
Greater than 80% as determined by SDS-PAGE. |