missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of G-protein signaling 20 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 20, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6604-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273602.1 | |
Regulator of G-protein signaling 20 Protein | |
Research Use Only | |
RGS20 | |
Recombinant Protein | |
E. coli | |
EPAGASSPAG RVDGGLQMGS ERMEMRKRQM PAAQDTPGAA PGQPGAGSRG SNACCFCWCC CCSCSCLTVR NQEDQRPTIA SHELRADLPT WEESPAPTLE EVNAWAQSFD KLMVTPAGRN AFREFLRTEF SEENMLFWMA CEELKKEANK NIIEEKARII YEDYISILSP KEVSLDSRVR EVINRNMVEP SQHIFDDAQL QIYTLMHRDS YPRFMNSAVY KDLLQSLSEK SIEA | |
Greater than 80% as determined by SDS-PAGE. |
8601 | |
100 μg | |
Not determined. | |
Full Length of BC015614 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |