missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human REG1B Protein

A cDNA sequence encoding the REG1B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13293-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943109

  • £4295.00 / 1mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

REG1B Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS HMQESQTELP NPRISCPEGT NAYRSYCYYF NEDPETWVDA DLYCQNMNSG NLVSVLTQAE GAFVASLIKE SSTDDSNVWI GLHDPKKNRR WHWSSGSLVS YKSWDTGSPS SANAGYCASL TSCSGFKKWK DESCEKKFSF VCKFKN
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 20mM Tris, pH 8.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers