missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RCHY1 Protein
A DNA sequence encoding the Homo sapiens (Human) RCHY1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7847-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009922.1 | |
RCHY1 Protein | |
Research Use Only | |
RCHY1 | |
Recombinant Protein | |
E. coli | |
MAATAREDGA SGQERGQRGC EHYDRGCLLK APCCDKLYTC RLCHDNNEDH QLDRFKVKEV QCINCEKIQH AQQTCEECST LFGEYYCDIC HLFDKDKKQY HCENCGICRI GPKEDFFHCL KCNLCLAMNL QGRHKCIENV SRQNCPICLE DIHTSRVVAH VLPCGHLLHR TCYEEMLKEG YRCPLCMHSA LDMTRYWRQL DDEVAQTPMP SEYQNMTVDI LCNDCNGRST VQFHILGMKC KICESYNTAQ AGGRRISLDQ Q | |
Greater than 80% as determined by SDS-PAGE. |
25898 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |