missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-5B Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6572-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001238965.1 | |
Ras-related protein Rab-5B Protein | |
Research Use Only | |
RAB5B | |
Recombinant Protein | |
E. coli | |
TSRSTARPNG QPQASKICQF KLVLLGESAV GKSSLVLRFV KGQFHEYQES TIGAAFLTQS VCLDDTTVKF EIWDTAGQER YHSLAPMYYR GAQAAIVVYD ITNQETFARA KTWVKELQRQ ASPSIVIALA GNKADLANKR MVEYEEAQAY ADDNSLLFME TSAKTAMNVN DLFLAIAKKL PKSEPQNLGG AAGRSRGVDL HEQSQQNKSQ CCSN | |
Greater than 80% as determined by SDS-PAGE. |
5869 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |