missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAP2A Protein
A cDNA sequence encoding the RAP2A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13267-2ug
Additional Details : Weight : 0.01000kg
Specifications
RAP2A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MREYKVVVLG SGGVGKSALT VQFVTGTFIE KYDPTIEDFY RKEIEVDSSP SVLEILDTAG TEQFASMRDL YIKNGQGFIL VYSLVNQQSF QDIKPMRDQI IRVKRYEKVP VILVGNKVDL ESEREVSSSE GRALAEEWGC PFMETSAKSK TMVDELFAEI VRQMNYAAQP DKDDPCCSAC | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAP2A solution (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |