missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RANBP1 Protein
A cDNA sequence encoding the RANBP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13261-1mg
Additional Details : Weight : 0.01000kg
Specifications
RANBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAAAKD THEDHDTSTE NTDESNHDPQ FEPIVSLPEQ EIKTLEEDEE ELFKMRAKLF RFASENDLPE WKERGTGDVK LLKHKEKGAI RLLMRRDKTL KICANHYITP MMELKPNAGS DRAWVWNTHA DFADECPKPE LLAIRFLNAE NAQKFKTKFE ECRKEIEERE KKAGSGKNDH AEKVAEKLEA LSVKEETKED AEEKQ | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RANBP1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol. |