missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB34 Protein
A cDNA sequence encoding the RAB34 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | RAB34 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952939
|
enQuireBio™
QP13238-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932939
|
enQuireBio™
QP13238-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942939
|
enQuireBio™
QP13238-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RAB34 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB34 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 40% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNILAPVRRD RVLAELPQCL RKEAALHGHK DFHPRVTCAC QEHRTGTVGF KISKVIVVGD LSVGKTCLIN RFCKDTFDKN YKATIGVDFE MERFEVLGIP FSLQLWDTAG QERFKCIAST YYRGAQAIII VFNLNDVASL EHTKQWLADA LKENDPSSVL LFLVGSKKDL STPAQYALME KDALQVAQEM KAEYWAVSSL TGENVREFFF RVAALTFEAN VLAELEKSGA RRIGDVVRIN SDDSNLYLTA SKKKPTCCP | |
Greater than 90% as determined by SDS-PAGE. |