missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB31 / Rab22B Protein

A DNA sequence encoding the Homo sapiens (Human) RAB31 / Rab22B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7541-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049270

  • £384.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006859.2
RAB31 / Rab22B Protein
Research Use Only
RAB31
Recombinant Protein
E. coli
MAIRELKVCL LGDTGVGKSS IVCRFVQDHF DHNISPTIGA SFMTKTVPCG NELHKFLIWD TAGQERFHSL APMYYRGSAA AVIVYDITKQ DSFYTLKKWV KELKEHGPEN IVMAIAGNKC DLSDIREVPL KDAKEYAESI GAIVVETSAK NAINIEELFQ GISRQIPPLD PHENGNNGTI KVEKPTMQAS RRCC
Greater than 80% as determined by SDS-PAGE.
11031
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers