missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB2A Protein

A DNA sequence encoding the Homo sapiens (Human) RAB2A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6569-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004240

  • £1688.38 / 1mg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001229573.1
RAB2A Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5862
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RAB2A
Recombinant Protein
E. coli
AYAYLFKYII IGDTGVGKSC LLLQFTDKRF QPVHDLTIGV EFGARMITID GKQIKLQIWD TAGQESFRSI TRSYYRGAAG ALLVYDITRR DTFNHLTTWL EDARQHSNSN MVIMLIGNKS DLESRREVKK EEGEAFAREH GLIFMETSAK TASNVEEAFI NTAKEIYEKI QEGVFDINNE ANGIKIGPQH AATNATHAGN QGGQQAGGGC C
Videos
Documents