missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAB1B Protein

A cDNA sequence encoding the RAB1B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13231-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942919

  • £99.15 / 2µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RAB1B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MNPEYDYLFK LLLIGDSGVG KSCLLLRFAD DTYTESYIST IGVDFKIRTI ELDGKTIKLQ IWDTAGQERF RTITSSYYRG AHGIIVVYDV TDQESYANVK QWLQEIDRYA SENVNKLLVG NKSDLTTKKV VDNTTAKEFA DSLGIPFLET SAKNATNVEQ AFMTMAAEIK KRMGPGAASG GERPNLKIDS TPVKPAGGGC C
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RAB1B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers