missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Putative 40S ribosomal protein S10-like Protein

A DNA sequence encoding the Homo sapiens (Human) Putative 40S ribosomal protein S10-like, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8151-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002931

  • £613.01 / 200µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

Putative 40S ribosomal protein S10-like Protein
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPS10P5
Recombinant Protein
E. coli
MLMPKKNRIA IHELLFKEGV MVAKKDVHMP KHPELADKNV PNLHVMKAMQ SLKSRGCVKE QFAWRHFYWY LTNEGSQYLR DYLHLPPEIV PATLHLPPEI VPATLHRSRP ETGRPRPKGL EGKRPARLTR READRDTYRR CSVPPGADKK AEAGAGSATE FQFRGRCGRG RGQPPQ
Videos
Documents