missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPS Protein
A cDNA sequence encoding the PTPS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13213-1mg
Additional Details : Weight : 0.01000kg
Specifications
PTPS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSTEGGGRRC QAQVSRRISF SASHRLYSKF LSDEENLKLF GKCNNPNGHG HNYKVVVTVH GEIDPATGMV MNLADLKKYM EEAIMQPLDH KNLDMDVPYF ADVVSTTENV AVYIWDNLQK VLPVGVLYKV KVYETDNNIV VYKGE | |
Greater than 95.0% as determined by Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
6-PyruvoylTetrahydropterin Synthase is formulated in 20mM Tris-HCl buffer pH 8.0, 1mM DTT and 20% glycerol. |