missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSTPIP1 Protein

A cDNA sequence encoding the PSTPIP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13198-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942809

  • £99.15 / 5µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PSTPIP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMMPQLQF KDAFWCRDFT AHTGYEVLLQ RLLDGRKMCK DMEELLRQRA QAEERYGKEL VQIARKAGGQ TEINSLRASF DSLKQQMENV GSSHIQLALT LREELRSLEE FRERQKEQRK KYEAVMDRVQ KSKLSLYKKA MESKKTYEQK CRDADDAEQA FERISANGHQ KQVEKSQNKA RQCKDSATEA ERVYRQSIAQ LEKVRAEWEQ EHRTTCEAFQ LQEFDRLTIL RNALWVHSNQ LSMQCVKDDE LYEEVRLTLE GCSIDADIDS FIQAKSTGTE PPAPVPYQNY YDREVTPLTS SPGIQPSCGM IKRFSGLLHG SPKTTSLAAS AASTETLTPT PERNEGVYTA IAVQEIQGNP ASPAQEYRAL YDYTAQNPDE LDLSAGDILE VILEGEDGWW TVERNGQRGF VPGSYLEKL
Greater than 90.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PSTPIP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers