missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSTPIP1 Protein
A cDNA sequence encoding the PSTPIP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13198-20ug
Additional Details : Weight : 0.01000kg
Specifications
PSTPIP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMPQLQF KDAFWCRDFT AHTGYEVLLQ RLLDGRKMCK DMEELLRQRA QAEERYGKEL VQIARKAGGQ TEINSLRASF DSLKQQMENV GSSHIQLALT LREELRSLEE FRERQKEQRK KYEAVMDRVQ KSKLSLYKKA MESKKTYEQK CRDADDAEQA FERISANGHQ KQVEKSQNKA RQCKDSATEA ERVYRQSIAQ LEKVRAEWEQ EHRTTCEAFQ LQEFDRLTIL RNALWVHSNQ LSMQCVKDDE LYEEVRLTLE GCSIDADIDS FIQAKSTGTE PPAPVPYQNY YDREVTPLTS SPGIQPSCGM IKRFSGLLHG SPKTTSLAAS AASTETLTPT PERNEGVYTA IAVQEIQGNP ASPAQEYRAL YDYTAQNPDE LDLSAGDILE VILEGEDGWW TVERNGQRGF VPGSYLEKL | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSTPIP1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |