missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMB6 Protein
A cDNA sequence encoding the PSMB6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13188-2ug
Additional Details : Weight : 0.01000kg
Specifications
PSMB6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTTIMAVQFD GGVVLGADSR TTTGSYIANR VTDKLTPIHD RIFCCRSGSA ADTQAVADAV TYQLGFHSIE LNEPPLVHTA ASLFKEMCYR YREDLMAGII IAGWDPQEGG QVYSVPMGGM MVRQSFAIGG SGSSYIYGYV DATYREGMTK EECLQFTANA LALAMERDGS SGGVIRLAAI AESGVERQVL LGDQIPKFAV ATLPPA | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMB6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.15M NaCl. |