missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein unc-119 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) Protein unc-119 homolog A, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001317095.1 |
---|---|
Gene ID (Entrez) | 9094 |
Name | Protein unc-119 homolog A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029620
|
enQuireBio™
QP7593-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049620
|
enQuireBio™
QP7593-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039620
|
enQuireBio™
QP7593-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059620
|
enQuireBio™
QP7593-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069620
|
enQuireBio™
QP7593-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079620
|
enQuireBio™
QP7593-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001317095.1 | |
Protein unc-119 homolog A Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9094 | |
Research Use Only | |
UNC119 | |
Recombinant Protein | |
E. coli | |
MKVKKGGGGA GTATESAPGP SGQSVAPIPQ PPAESESGSE SEPDAGPGPR PGPLQRKQPI GPEDVLGLQR ITGDYLCSPE ENIYKIDFVR FKIRDMDSGT VLFEIKKPPV SERLPINRRD LDPNAGRFVR YQFTPAFLRL RQVGATVEFT VGDKPVNNFR MIERHYFRNQ LLKSFDFHFG FCIPSSKNTC EHIYDFPPLS EELISEMIRH PYETQSDSFY FVDDRLVMHN KADYSYSGTP | |
Greater than 90% as determined by SDS-PAGE. |