missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein FAM49B Protein
A DNA sequence encoding the Homo sapiens (Human) Protein FAM49B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8121-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243692.1 | |
Protein FAM49B Protein | |
Research Use Only | |
FAM49B | |
Recombinant Protein | |
E. coli | |
MGNLLKVLTC TDLEQGPNFF LDFENAQPTE SEKEIYNQVN VVLKDAEGIL EDLQSYRGAG HEIREAIQHP ADEKLQEKAW GAVVPLVGKL KKFYEFSQRL EAALRGLLGA LTSTPYSPTQ HLEREQALAK QFAEILHFTL RFDELKMTNP AIQNDFSYYR RTLSRMRINN VPAEGENEVN NELANRMSLF YAEATPMLKT LSDATTKFVS ENKNLPIENT TDCLSTMASV CRVMLETPEY RSRFTNEETV SFCLRVMVGV IILYDHVHPV GAFAKTSKID MKGCIKVLKD QPPNSVEGLL NALRYTTKHL NDETTSKQIK SMLQ | |
Greater than 90% as determined by SDS-PAGE. |
51571 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |