missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein chibby homolog 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein chibby homolog 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8286-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003781

  • £145.46 / 10µg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001002880.2
Protein chibby homolog 1 Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
25776
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CBY1
Recombinant Protein
E. coli
MPFFGNTFSP KKTPPRKSAS LSNLHSLDRS TREVELGLEY GSPTMNLAGQ SLKFENGQWI AETGVSGGVD RREVQRLRRR NQQLEEENNL LRLKVDILLD MLSESTAESH LMEKELDELR ISRKRK
Videos
Documents