missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein arginine N-methyltransferase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein arginine N-methyltransferase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8434-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001527.3 | |
Protein arginine N-methyltransferase 1 Protein | |
Research Use Only | |
PRMT1 | |
Recombinant Protein | |
E. coli | |
MENFVATLAN GMSLQPPLEE VSCGQAESSE KPNAEDMTSK DYYFDSYAHF GIHEEMLKDE VRTLTYRNSM FHNRHLFKDK VVLDVGSGTG ILCMFAAKAG ARKVIGIECS SISDYAVKIV KANKLDHVVT IIKGKVEEVE LPVEKVDIII SEWMGYCLFY ESMLNTVLYA RDKWLAPDGL IFPDRATLYV TAIEDRQYKD YKIHWWENVY GFDMSCIKDV AIKEPLVDVV DPKQLVTNAC LIKEVDIY | |
Greater than 90% as determined by SDS-PAGE. |
3276 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |