missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PROSC Protein
A cDNA sequence encoding the PROSC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13158-1mg
Additional Details : Weight : 0.01000kg
Specifications
PROSC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMWRAGSM SAELGVGCAL RAVNERVQQA VARRPRDLPA IQPRLVAVSK TKPADMVIEA YGHGQRTFGE NYVQELLEKA SNPKILSLCP EIKWHFIGHL QKQNVNKLMA VPNLFMLETV DSVKLADKVN SSWQRKGSPE RLKVMVQINT SGEESKHGLP PSETIAIVEH INAKCPNLEF VGLMTIGSFG HDLSQGPNPD FQLLLSLREE LCKKLNIPAD QVELSMGMSA DFQHAVEVGS TNVRIGSTIF GERDYSKKPT PDKCAADVKA PLEVAQEH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PROSC solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |