missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proguanylin Protein
A cDNA sequence encoding the Proguanylin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13157-1mg
Additional Details : Weight : 0.01000kg
Specifications
Proguanylin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS VTVQDGNFSF SLESVKKLKD LQEPQEPRVG KLRNFAPIPG EPVVPILCSN PNFPEELKPL CKEPNAQEIL QRLEEIAEDP GTCEICAYAA CTGC | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Proguanylin filtered (0.4 microm) and lyophilized in 0.5 mg/ml in deionized H20. |