missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRAME Protein
A DNA sequence encoding the Homo sapiens (Human) PRAME, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6524-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001278644.1 | |
PRAME Protein | |
Research Use Only | |
PRAME | |
Recombinant Protein | |
E. coli | |
MERRRLWGSI QSRYISMSVW TSPRRLVELA GQSLLKDEAL AIAALELLPR ELFPPLFMAA FDGRHSQTLK AMVQAWPFTC LPLGVLMKGQ HLHLETFKAV LDGLDVLLAQ EVRPRRWKLQ VLDLRKNSHQ DFWTVWSGNR ASLYSFPEPE AAQPMTKKRK VDGLSTEAEQ PFIPVEVLVD LFLKEGACDE LFSYLIEKVK RKKNVLRLCC KKLKIFAMPM QDIKMILKMV QLDSIEDLEV TCTWKLPTLA KFSPYLGQMI NLRRLLLSHI HASSYISPEK EEQYIAQFTS QFLSLQCLQA LYVDSLFFLR GRLDQLLRHV MNPLETLSIT NCRLSEGDVM HLSQSPSVSQ LSVLSLSGVM LTDVSPEPLQ ALLERASATL QDLVFDECGI TDDQLLALLP SLSHCSQLTT LSFYGNSISI SALQSLLQHL IGLSNLTHVL YPVPLESYED IHGTLHLERL AYLHARLREL LCELGRPSMV WLSANPCPHC GDRTFYDPEP ILCPCFMPN | |
Greater than 90% as determined by SDS-PAGE. |
23532 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |