missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRAME Protein
A DNA sequence encoding the Homo sapiens (Human) PRAME, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001278644.1 |
---|---|
Gene ID (Entrez) | 23532 |
Name | PRAME Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083900
|
enQuireBio™
QP6524-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003910
|
enQuireBio™
QP6524-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093900
|
enQuireBio™
QP6524-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013910
|
enQuireBio™
QP6524-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023910
|
enQuireBio™
QP6524-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033910
|
enQuireBio™
QP6524-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001278644.1 | |
PRAME Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
23532 | |
Research Use Only | |
PRAME | |
Recombinant Protein | |
E. coli | |
MERRRLWGSI QSRYISMSVW TSPRRLVELA GQSLLKDEAL AIAALELLPR ELFPPLFMAA FDGRHSQTLK AMVQAWPFTC LPLGVLMKGQ HLHLETFKAV LDGLDVLLAQ EVRPRRWKLQ VLDLRKNSHQ DFWTVWSGNR ASLYSFPEPE AAQPMTKKRK VDGLSTEAEQ PFIPVEVLVD LFLKEGACDE LFSYLIEKVK RKKNVLRLCC KKLKIFAMPM QDIKMILKMV QLDSIEDLEV TCTWKLPTLA KFSPYLGQMI NLRRLLLSHI HASSYISPEK EEQYIAQFTS QFLSLQCLQA LYVDSLFFLR GRLDQLLRHV MNPLETLSIT NCRLSEGDVM HLSQSPSVSQ LSVLSLSGVM LTDVSPEPLQ ALLERASATL QDLVFDECGI TDDQLLALLP SLSHCSQLTT LSFYGNSISI SALQSLLQHL IGLSNLTHVL YPVPLESYED IHGTLHLERL AYLHARLREL LCELGRPSMV WLSANPCPHC GDRTFYDPEP ILCPCFMPN | |
Greater than 90% as determined by SDS-PAGE. |