missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP3R1 Protein
A cDNA sequence encoding the PPP3R1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13129-1mg
Additional Details : Weight : 0.01000kg
Specifications
PPP3R1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGNEASYPLE MCSHFDADEI KRLGKRFKKL DLDNSGSLSV EEFMSLPELQ QNPLVQRVID IFDTDGNGEV DFKEFIEGVS QFSVKGDKEQ KLRFAFRIYD MDKDGYISNG ELFQVLKMMV GNNLKDTQLQ QIVDKTIINA DKDGDGRISF EEFCAVVGGL DIHKKMVVDV | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PPP3R1 Human solution (1 mg/1ml) containing 20mM Trsi HCL pH 8.0, 0.1M NaCl, 2mM DTT and 10% glycerol. |