missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1R2 Protein
A cDNA sequence encoding the PPP1R2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13125-20ug
Additional Details : Weight : 0.01000kg
Specifications
PPP1R2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAASTAS HRPIKGILKN KTSTTSSMVA SAEQPRGNVD EELSKKSQKW DEMNILATYH PADKDYGLMK IDEPSTPYHS MMGDDEDACS DTEATEAMAP DILARKLAAA EGLEPKYRIQ EQESSGEEDS DLSPEEREKK RQFEMKRKLH YNEGLNIKLA RQLISKDLH | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PPP1R2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT. |