missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPP1CC Protein

A cDNA sequence encoding the PPP1CC was constructed and used to recombinantly synthesize the protein.

£98.39 - £1324.47

Specifications

Name PPP1CC Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15972569
View Documents Promotion Details
enQuireBio™
QP13122-2UG
2 μg
£98.39
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 01-03-2022
Please to purchase this item. Need a web account? Register with us today!
15962569
View Documents Promotion Details
enQuireBio™
QP13122-10UG
10 μg
£151.37
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 01-03-2022
Please to purchase this item. Need a web account? Register with us today!
15952569
View Documents Promotion Details
enQuireBio™
QP13122-100UG
100 μg
£1324.47
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 01-03-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PPP1CC Protein
His
Human
Greater than 85% as determined by SDS-PAGE.
PPP1CC
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MADLDKLNID SIIQRLLEVR GSKPGKNVQL QENEIRGLCL KSREIFLSQP ILLELEAPLK ICGDIHGQYY DLLRLFEYGG FPPESNYLFL GDYVDRGKQS LETICLLLAY KIKYPENFFL LRGNHECASI NRIYGFYDEC KRRYNIKLWK TFTDCFNCLP IAAIVDEKIF CCHGGLSPDL QSMEQIRRIM RPTDVPDQGL LCDLLWSDPD KDVLGWGEND RGVSFTFGAE VVAKFLHKHD LDLICRAHQV VEDGYEFFAK RQLVTLFSAP NYCGEFDNAG AMMSVDETLM CSFQILKPAE KKKPNATRPV TPPRGMITKQ AKK
5501
SDS
Product Certifications