missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPIL3 Protein
A cDNA sequence encoding the PPIL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10833-20ug
Additional Details : Weight : 0.01000kg
Specifications
53938 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 280 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
His | |
PPIL3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |
PPIL3 Protein | |
Research Use Only | |
PPIL3 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSVTLHTDVG DIKIEVFCER TPKTCENFLA LCASNYYNGC IFHRNIKGFM VQTGDPTGTG RGGNSIWGKK FEDEYSEYLK HNVRGVVSMA NNGPNTNGSQ FFITYGKQPH LDMKYTVFGK VIDGLETLDE LEKLPVNEKT YRPLNDVHIK DITIHANPFA Q | |
Greater than 90.0% as determined by SDS-PAGE. |