missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPIL3 Protein

A cDNA sequence encoding the PPIL3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10833-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955308

  • £99.15 / 5µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

53938
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 280 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
His
PPIL3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
PPIL3 Protein
Research Use Only
PPIL3
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSVTLHTDVG DIKIEVFCER TPKTCENFLA LCASNYYNGC IFHRNIKGFM VQTGDPTGTG RGGNSIWGKK FEDEYSEYLK HNVRGVVSMA NNGPNTNGSQ FFITYGKQPH LDMKYTVFGK VIDGLETLDE LEKLPVNEKT YRPLNDVHIK DITIHANPFA Q
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers