missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POU6F1 Protein

A cDNA sequence encoding the POU6F1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13111-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942539

  • £2622.00 / 1mg

Estimated Shipment
29-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

POU6F1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The POU5F1 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPGISSQ ILTNAQGQVI GTLPWVVNSA SVAAPAPAQS LQVQAVTPQL LLNAQGQVIA TLASSPLPPP VAVRKPSTPE SPAKSEVQPI QPTPTVPQPA VVIASPAPAA KPSASAPIPI TCSETPTVSQ LVSKPHTPSL DEDGINLEEI REFAKNFKIR RLSLGLTQTQ VGQALTATEG PAYSQSAICR FEKLDITPKS AQKLKPVLEK WLNEAELRNQ EGQQNLMEFV GGEPSKKRKR RTSFTPQAIE ALNAYFEKNP LPTGQEITEI AKELNYDREV VRVWFCNRRQ TLKNTSKLNV FQIP
Videos
SDS
Documents