missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2I Protein
A cDNA sequence encoding the POLR2I was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13101-2ug
Additional Details : Weight : 0.01000kg
Specifications
POLR2I Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPDGTY EPGFVGIRFC QECNNMLYPK EDKENRILLY ACRNCDYQQE ADNSCIYVNK ITHEVDELTQ IIADVSQDPT LPRTEDHPCQ KCGHKEAVFF QSHSARAEDA MRLYYVCTAP HCGHRWTE | |
Greater than 85% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
POLR2I protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT. |