missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2E Protein
A cDNA sequence encoding the POLR2E was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | POLR2E Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15972499
|
enQuireBio™
QP13098-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962499
|
enQuireBio™
QP13098-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952499
|
enQuireBio™
QP13098-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
POLR2E Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLR2E solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDEEET YRLWKIRKTI MQLCHDRGYL VTQDELDQTL EEFKAQFGDK PSEGRPRRTD LTVLVAHNDD PTDQMFVFFP EEPKVGIKTI KVYCQRMQEE NITRALIVVQ QGMTPSAKQS LVDMAPKYIL EQFLQQELLI NITEHELVPE HVVMTKEEVTELLARYKLRE NQLPRIQAGD PVARYFGIKR GQVVKIIRPS ETAGRYITYR LVQ | |
Greater than 90% as determined by SDS-PAGE. |